Learn More
Waters Corp Waters BioResolve RP mAb Polyphenyl Kit, 450Å, 2.7 μm
Supplier:  Waters Corp 186008961
Encompass_Preferred
This BioResolve RP mAb Polyphenyl Method Validation Kit is designed for reversed-phase protein separation and analysis of intact IgG monoclonal antibodies and fragments, containing three columns for method validation in biotherapeutic drug analysis.
  • Reversed phase
  • 90°C Temp
  • 10000 psi (689 Bar) Pressure
  • 2.7 μm Particle
  • 450 Å Pore
  • Protein QC
  • HPLC, UHPLC, UPLC compatible
  • L11 USP
  • 3 Units
  • Intact protein, mAb subunit, ADC, AAV analysis
Catalog No. 50-152-8601