Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Method Validation Kit, BioResolve Polyphenyl, 450Å, 2.7 μm

Catalog No. 501528577 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The BioResolve RP mAb Method Validation Kit offers three columns with 2.7μm superficially-porous-silica particles and 450Å pore diameter. Ideal for LC or LC-MS analyses of intact IgG monoclonal antibodies and fragments, the kit includes columns from different batches, along with Intact mAb Mass Check Standard and mAb Subunit Standard for performance benchmarking, ensuring robust performance.

  • Separation mode: Reversed Phase
  • Particle size: 2.7 μm
  • Pore size: 450 Å
  • Column kits
  • 3 units per package
  • Column dimensions: 4.6 mm × 100 mm
  • Superficially-porous silica particle composition
  • Proprietary polyphenyl bonding process
  • Application for LC or LC-MS analysis of intact IgG monoclonal antibodies and fragments
  • Included standards: Intact mAb Mass Check Standard and mAb Subunit Standard

Catalog No. 50-152-8577 Supplier Waters Corp Supplier No. 176004170
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The BioResolve RP mAb Method Validation Kit offers three columns with 2.7μm superficially-porous-silica particles and 450Å pore diameter. Ideal for LC or LC-MS analyses of intact IgG monoclonal antibodies and fragments, the kit includes columns from different batches, along with Intact mAb Mass Check Standard and mAb Subunit Standard for performance benchmarking, ensuring robust performance.

  • Separation mode: Reversed Phase
  • Particle size: 2.7 μm
  • Pore size: 450 Å
  • Column kits
  • 3 units per package
  • Column dimensions: 4.6 mm × 100 mm
  • Superficially-porous silica particle composition
  • Proprietary polyphenyl bonding process
  • Application for LC or LC-MS analysis of intact IgG monoclonal antibodies and fragments
  • Included standards: Intact mAb Mass Check Standard and mAb Subunit Standard

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.