Learn More
Waters Corp Waters VanGuard Cartridge, 450Å, 2.7 μm, 2.1 mm X 5 mm, 3 pk
Catalog No. 501528579 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
BioResolve RP mAb Columns contain 2.7-μm superficially-porous-silica particles with an average pore diameter of 450Å. These particles are functionalized with a proprietary polyphenyl bonding process designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments. This product acts as an all-in-one column protection solution by removing contaminants from the analytical workflow.
  • Designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments.
  • Removes contaminants from the analytical workflow.
  • Filters out contaminants to prevent them from negatively affecting the analytical column and data.
  • Reusable, and a worn one can be replaced.
  • Offers the same inner diameter and pH range as the corresponding polyphenyl column, ensuring chemical and mechanical stability.
Catalog No. 50-152-8579 Supplier Waters Corp Supplier No. 176004212
BioResolve RP mAb Columns contain 2.7-μm superficially-porous-silica particles with an average pore diameter of 450Å. These particles are functionalized with a proprietary polyphenyl bonding process designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments. This product acts as an all-in-one column protection solution by removing contaminants from the analytical workflow.
  • Designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments.
  • Removes contaminants from the analytical workflow.
  • Filters out contaminants to prevent them from negatively affecting the analytical column and data.
  • Reusable, and a worn one can be replaced.
  • Offers the same inner diameter and pH range as the corresponding polyphenyl column, ensuring chemical and mechanical stability.