Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters VanGuard Cartridge, Polyphenyl, 450Å, 2.7 µm, 2.1 × 5 mm

Catalog No. 501528590 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These cartridges utilize 2.7 µm superficially-porous-silica particles with a 450Å pore diameter and polyphenyl bonding. Designed for LC or LC-MS analysis of intact IgG monoclonal antibodies and fragments, they prevent column contamination and integrate into workflows, extending column lifetimes.

  • Separation mode: reversed phase.
  • Temperature limits: 90°C.
  • Maximum pressure: 10000 psi (689 bar).
  • Particle size: 2.7 µm; Pore: 450Å.
  • QC tested for protein.
  • System compatibility: HPLC, UHPLC, UPLC.
  • USP classification: L11.
  • Package contains 3 units.
  • Dimensions: 2.1 × 5 mm.
  • Material: superficially-porous-silica with polyphenyl bonding.
  • Prevents column contamination; enhances chemical and mechanical stability.
  • Improves column lifetimes.
  • Requires universal cartridge holder.

Catalog No. 50-152-8590 Supplier Waters Corp Supplier No. 186008943
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These cartridges utilize 2.7 µm superficially-porous-silica particles with a 450Å pore diameter and polyphenyl bonding. Designed for LC or LC-MS analysis of intact IgG monoclonal antibodies and fragments, they prevent column contamination and integrate into workflows, extending column lifetimes.

  • Separation mode: reversed phase.
  • Temperature limits: 90°C.
  • Maximum pressure: 10000 psi (689 bar).
  • Particle size: 2.7 µm; Pore: 450Å.
  • QC tested for protein.
  • System compatibility: HPLC, UHPLC, UPLC.
  • USP classification: L11.
  • Package contains 3 units.
  • Dimensions: 2.1 × 5 mm.
  • Material: superficially-porous-silica with polyphenyl bonding.
  • Prevents column contamination; enhances chemical and mechanical stability.
  • Improves column lifetimes.
  • Requires universal cartridge holder.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.