Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters VanGuard Cartridge, Polyphenyl, 450Å, 2.7 μm, 3.9 mm × 5 mm

Catalog No. 501528594 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

  • Enhances analytical column robustness and lifetime.
  • Features 2.7 μm superficially porous silica particles.
  • Pore size: 450 Å.
  • Proprietary polyphenyl bonding.
  • Optimized for reversed-phase LC/LC-MS of intact IgG monoclonal antibodies and fragments.
  • Guard column traps contaminants.
  • Separation mode: Reversed Phase.
  • Temperature limit: 90 °C.
  • Max pressure: 10000 psi (689 Bar).
  • QC tested: Protein.
  • System compatibility: HPLC, UHPLC, UPLC.
  • USP Classification: L11.
  • Dimensions: 3.9 mm × 5 mm.
  • Package: Three cartridges.

Catalog No. 50-152-8594 Supplier Waters Corp Supplier No. 186008947
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Enhances analytical column robustness and lifetime.
  • Features 2.7 μm superficially porous silica particles.
  • Pore size: 450 Å.
  • Proprietary polyphenyl bonding.
  • Optimized for reversed-phase LC/LC-MS of intact IgG monoclonal antibodies and fragments.
  • Guard column traps contaminants.
  • Separation mode: Reversed Phase.
  • Temperature limit: 90 °C.
  • Max pressure: 10000 psi (689 Bar).
  • QC tested: Protein.
  • System compatibility: HPLC, UHPLC, UPLC.
  • USP Classification: L11.
  • Dimensions: 3.9 mm × 5 mm.
  • Package: Three cartridges.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.