Learn More
Waters Corp BioResolve RP mAb Polyphenyl VanGuard Cartridge w/holder, 450Å, 2.7 µm, 3.9 mm X 5 mm, 3/pk
Catalog No. 501528573 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
BioResolve RP mAb Columns contain 2.7-μm superficially-porous-silica particles having an average pore diameter of 450Å. The particles are functionalized with a proprietary polyphenyl bonding process that was specifically designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments. This Part Number contains (3) VanGuard Cartridges as well as an included VanGuard Universal Cartridge Holder (P/N 186007949).
Catalog No. 50-152-8573 Supplier Waters Corp Supplier No. 176004161
BioResolve RP mAb Columns contain 2.7-μm superficially-porous-silica particles having an average pore diameter of 450Å. The particles are functionalized with a proprietary polyphenyl bonding process that was specifically designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments. This Part Number contains (3) VanGuard Cartridges as well as an included VanGuard Universal Cartridge Holder (P/N 186007949).