Learn More
Waters Corp Waters BioResolve SCX mAb Column, 3 μm, 2.1 mm, 100 mm
Supplier:  Waters Corp 186009056
Encompass_Preferred
The BioResolve SCX mAb Column is a corrosion-resistant column with non-porous, polymer-based particles and negatively charged sulfonic acid ligands, designed for high-resolution, charge-based separations of monoclonal antibodies and other proteins in LC and LC-MS applications.
  • Corrosion-resistant column with non-porous, polymer-based particles.
  • Provides reproducible, high-resolution, charge-based separations of monoclonal antibodies and other proteins.
  • Suitable for LC and LC-MS applications using robust salt or pH gradient methods.
  • QC tested to ensure consistent performance.
  • Compatible with UHPLC, UPLC, and HPLC systems, offering enhanced sample loading.
  • Designed for charge variant analysis of monoclonal antibodies, with optimal diffusion kinetics and pressure tolerance.
  • Includes a free vial of mAb charge variant standard for performance testing.
Catalog No. 50-203-8890