Learn More
Waters Corp BioResolve SCX mAb Column, 3 μm, 4.6 × 50 mm, VanGuard FIT
Catalog No. 502392646 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The BioResolve SCX mAb Column with VanGuard FIT is a corrosion-resistant column containing non-porous, polymer-based particles grafted with a multi-component network of negatively charged sulfonic acid ligands. It delivers reproducible, high-resolution, charged-based separations of monoclonal antibodies and other proteins in LC and LC-MS applications using robust salt or pH gradient methods.
  • Optimized for SCX analysis with a multi-component network of negatively charged sulfonic acid ligands.
  • Delivers reproducible, high-resolution separations for accurate charge-variant quantitation.
  • Features VanGuard Fully Integrated Technology (FIT) for enhanced column life and protection from contamination.
  • Each batch is QC tested to ensure consistent performance.
  • Includes eCord for connectivity.
Catalog No. 50-239-2646 Supplier Waters Corp Supplier No. 186009057
The BioResolve SCX mAb Column with VanGuard FIT is a corrosion-resistant column containing non-porous, polymer-based particles grafted with a multi-component network of negatively charged sulfonic acid ligands. It delivers reproducible, high-resolution, charged-based separations of monoclonal antibodies and other proteins in LC and LC-MS applications using robust salt or pH gradient methods.
  • Optimized for SCX analysis with a multi-component network of negatively charged sulfonic acid ligands.
  • Delivers reproducible, high-resolution separations for accurate charge-variant quantitation.
  • Features VanGuard Fully Integrated Technology (FIT) for enhanced column life and protection from contamination.
  • Each batch is QC tested to ensure consistent performance.
  • Includes eCord for connectivity.