Learn More
Waters Corp BioResolve SCX mAb Column, 3 μm, 2.1 x 50 mm, with VanGuard FIT
Catalog No. 502393015 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The BioResolve SCX mAb column is designed for high-resolution, charge-based separations of monoclonal antibodies and other proteins. It utilizes 3 μm, non-porous particles with finely tuned surface chemistry for unprecedented resolution and is suitable for both LC and LC-MS applications.
  • High resolution: Utilizes 3 μm, non-porous particles with finely tuned surface chemistry and sulfonic acid grafting for unprecedented resolution.
  • Reproducibility: Optimized for reproducible, high-resolution, charge-based separations of mAbs and other proteins.
  • Flexibility: Suitable for both LC and LC-MS applications using robust salt or pH gradient methods.
  • Column protection: Can be integrated with VanGuard™ FIT (Fully Integrated Technology) for no-compromise column protection and extended lifetimes.
Catalog No. 50-239-3015 Supplier Waters Corp Supplier No. 176004341
The BioResolve SCX mAb column is designed for high-resolution, charge-based separations of monoclonal antibodies and other proteins. It utilizes 3 μm, non-porous particles with finely tuned surface chemistry for unprecedented resolution and is suitable for both LC and LC-MS applications.
  • High resolution: Utilizes 3 μm, non-porous particles with finely tuned surface chemistry and sulfonic acid grafting for unprecedented resolution.
  • Reproducibility: Optimized for reproducible, high-resolution, charge-based separations of mAbs and other proteins.
  • Flexibility: Suitable for both LC and LC-MS applications using robust salt or pH gradient methods.
  • Column protection: Can be integrated with VanGuard™ FIT (Fully Integrated Technology) for no-compromise column protection and extended lifetimes.