Learn More
Waters Corp HPLC Column, BioResolve SCX mAb, 3 μm, 4.6 mm × 100 mm, VanGuard FIT
Catalog No. 502393024 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
BioResolve SCX mAb, 3 μm, corrosion-resistant columns utilize non-porous, polymer-based particles grafted with a multi-component network of negatively charged sulfonic acid ligands. This technology delivers reproducible, high-resolution, charged-based separations of mAbs and other proteins in LC and LC-MS applications using robust salt or pH gradient methods.
  • Suitable for both LC and LC-MS applications.
  • Supports robust salt or pH gradient methods.
  • Includes VanGuard™ fully integrated technology (FIT) to maximize column life without degrading biomolecule component resolution.
  • Operates within a pH range of 3 to 11.
Catalog No. 50-239-3024 Supplier Waters Corp Supplier No. 176004348
BioResolve SCX mAb, 3 μm, corrosion-resistant columns utilize non-porous, polymer-based particles grafted with a multi-component network of negatively charged sulfonic acid ligands. This technology delivers reproducible, high-resolution, charged-based separations of mAbs and other proteins in LC and LC-MS applications using robust salt or pH gradient methods.
  • Suitable for both LC and LC-MS applications.
  • Supports robust salt or pH gradient methods.
  • Includes VanGuard™ fully integrated technology (FIT) to maximize column life without degrading biomolecule component resolution.
  • Operates within a pH range of 3 to 11.