Learn More
Waters Corp BioResolve SCX mAb Column, VanGuard FIT, 3 μm, 4.6 mm x 50 mm
Catalog No. 502392873 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The BioResolve SCX mAb, 3 μm, corrosion–resistant columns use non–porous, polymer-based particles with sulfonic acid ligands for high–resolution, charged-based separations of mAbs and other proteins in LC/LC-MS. Its SCX stationary phase ensures excellent separation efficiency. Includes VanGuard™ Fully Integrated Technology (FIT) for extended column life and a mAb Charge Variant Standard.
  • High-performance liquid chromatography for mAb characterization.
  • Exceptional separation efficiency and resolution for mAbs.
  • Versatile for mAb purity, stability testing, and drug discovery.
  • Compatible with a wide range of mobile phases.
  • VanGuard™ Fully Integrated Technology (FIT) protects against contamination; extends column life.
  • Includes mAb Charge Variant Standard for performance verification.
  • User-friendly design and easy maintenance.
Catalog No. 50-239-2873 Supplier Waters Corp Supplier No. 176004346
The BioResolve SCX mAb, 3 μm, corrosion–resistant columns use non–porous, polymer-based particles with sulfonic acid ligands for high–resolution, charged-based separations of mAbs and other proteins in LC/LC-MS. Its SCX stationary phase ensures excellent separation efficiency. Includes VanGuard™ Fully Integrated Technology (FIT) for extended column life and a mAb Charge Variant Standard.
  • High-performance liquid chromatography for mAb characterization.
  • Exceptional separation efficiency and resolution for mAbs.
  • Versatile for mAb purity, stability testing, and drug discovery.
  • Compatible with a wide range of mobile phases.
  • VanGuard™ Fully Integrated Technology (FIT) protects against contamination; extends column life.
  • Includes mAb Charge Variant Standard for performance verification.
  • User-friendly design and easy maintenance.