Learn More
Waters Corp BioResolve SCX mAb Column, 3 μm, 2.1 mm x 100 mm
Catalog No. 502038889 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The BioResolve SCX mAb Column with VanGuard FIT is a corrosion-resistant column featuring non-porous, polymer-based particles with optimized sulfonic acid ligands. It delivers reproducible, high-resolution, charge-based separations of monoclonal antibodies (mAbs) and other proteins in LC and LC-MS applications. With VanGuard FIT, it maximizes column life, and each batch is QC tested for consistent performance. It is designed for charge variant analysis of monoclonal antibodies.
  • Corrosion-resistant design.
  • Non-porous, polymer-based particles with optimized ligands.
  • Reproducible, high-resolution separations for mAbs and proteins.
  • Compatible with LC and LC-MS.
  • VanGuard FIT for extended column life and maintained resolution.
  • Batch QC tested for consistent performance.
  • Designed for charge variant analysis of monoclonal antibodies.
Catalog No. 50-203-8889 Supplier Waters Corp Supplier No. 186009055
The BioResolve SCX mAb Column with VanGuard FIT is a corrosion-resistant column featuring non-porous, polymer-based particles with optimized sulfonic acid ligands. It delivers reproducible, high-resolution, charge-based separations of monoclonal antibodies (mAbs) and other proteins in LC and LC-MS applications. With VanGuard FIT, it maximizes column life, and each batch is QC tested for consistent performance. It is designed for charge variant analysis of monoclonal antibodies.
  • Corrosion-resistant design.
  • Non-porous, polymer-based particles with optimized ligands.
  • Reproducible, high-resolution separations for mAbs and proteins.
  • Compatible with LC and LC-MS.
  • VanGuard FIT for extended column life and maintained resolution.
  • Batch QC tested for consistent performance.
  • Designed for charge variant analysis of monoclonal antibodies.