Learn More
Waters Corp WATERS CORPORATION Column, SCX, 3 μm, 4.6 mm, 100 mm
Catalog No. 502038892 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The BioResolve SCX mAb Column with VanGuard FIT features corrosion-resistant, non-porous polymer particles with negatively charged sulfonic acid ligands. It enables reproducible, high-resolution, charged-based separations of mAbs and proteins in LC/LC-MS applications via salt or pH gradient methods.
  • Delivers reproducible, high-resolution, charged-based separations of mAbs and proteins in LC/LC-MS.
  • Each batch is QC tested; VanGuard FIT maximizes column life.
  • Suggested for charge variant analysis.
  • Increases peak capacity for better component resolution.
  • Manufactured in a cGMP, ISO 9001 certified facility.
  • Individually tested; Performance Chromatogram and Certificate of Batch Analysis with eCord™ Intelligent Chip.
  • Allows lower temperature studies, reducing on-column sample degradation.
Catalog No. 50-203-8892 Supplier Waters Corp Supplier No. 186009059
The BioResolve SCX mAb Column with VanGuard FIT features corrosion-resistant, non-porous polymer particles with negatively charged sulfonic acid ligands. It enables reproducible, high-resolution, charged-based separations of mAbs and proteins in LC/LC-MS applications via salt or pH gradient methods.
  • Delivers reproducible, high-resolution, charged-based separations of mAbs and proteins in LC/LC-MS.
  • Each batch is QC tested; VanGuard FIT maximizes column life.
  • Suggested for charge variant analysis.
  • Increases peak capacity for better component resolution.
  • Manufactured in a cGMP, ISO 9001 certified facility.
  • Individually tested; Performance Chromatogram and Certificate of Batch Analysis with eCord™ Intelligent Chip.
  • Allows lower temperature studies, reducing on-column sample degradation.