Learn More
Waters Corp BioResolve SCX mAb Kit, 3 μm, 4.6 x 100 mm
Catalog No. 502390775 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This BioResolve SCX mAb Method Validation Kit provides corrosion-resistant columns for reproducible, high-resolution, charge-based separations of monoclonal antibodies and other proteins. It includes three columns from different batches and a charge variant standard for performance verification.
  • Provides reproducible, high-resolution, charge-based separations.
  • Features corrosion-resistant columns with non-porous particles.
  • Includes three columns from different batches.
  • Supplied with a charge variant standard.
  • Compatible with LC, LC-MS, UHPLC, and UPLC applications.
  • Supports quick gradient preparation.
  • Utilizes advanced cation-exchange bonding technology.
  • Extends column lifetime with VanGuard FIT protection.
  • Minimizes band spread and enhances separation power.
  • Simplifies method transfer with sample loading features.
Catalog No. 50-239-0775 Supplier Waters Corp Supplier No. 186009200
Add to Cart
Add to Cart
Request a Quote
This BioResolve SCX mAb Method Validation Kit provides corrosion-resistant columns for reproducible, high-resolution, charge-based separations of monoclonal antibodies and other proteins. It includes three columns from different batches and a charge variant standard for performance verification.
  • Provides reproducible, high-resolution, charge-based separations.
  • Features corrosion-resistant columns with non-porous particles.
  • Includes three columns from different batches.
  • Supplied with a charge variant standard.
  • Compatible with LC, LC-MS, UHPLC, and UPLC applications.
  • Supports quick gradient preparation.
  • Utilizes advanced cation-exchange bonding technology.
  • Extends column lifetime with VanGuard FIT protection.
  • Minimizes band spread and enhances separation power.
  • Simplifies method transfer with sample loading features.