Learn More
Waters Corp BioResolve SCX mAb Method Validation Kit, 3 μm, 2.1 mm x 100 mm, 3 per pack
Catalog No. 502390780 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This HPLC Column Kit is designed for method validation using strong cation-exchange (SCX) chemistry. It features a 3 μm particle size and column dimensions of 2.1 mm x 100 mm. The kit includes 3 columns, suitable for analytical applications with pH stability from 3.0 to 11.0.
  • 3 μm particle size
  • 2.1 mm x 100 mm column dimensions
  • Kit contains 3 pieces
  • Strong cation-exchange chemistry
  • SCX mAb chemistry
  • USP Class L34
  • Analytical column type
  • Stainless steel hardware material
  • Ready-to-use column hardware
  • pH stability from 3.0 to 11.0
  • Ion exchange chromatography separation mode
  • HPLC technology
  • Polymer matrix
  • Spherical, non-porous particle morphology
  • Pressure stability up to 689 bar
Catalog No. 50-239-0780 Supplier Waters Corp Supplier No. 186009198
This HPLC Column Kit is designed for method validation using strong cation-exchange (SCX) chemistry. It features a 3 μm particle size and column dimensions of 2.1 mm x 100 mm. The kit includes 3 columns, suitable for analytical applications with pH stability from 3.0 to 11.0.
  • 3 μm particle size
  • 2.1 mm x 100 mm column dimensions
  • Kit contains 3 pieces
  • Strong cation-exchange chemistry
  • SCX mAb chemistry
  • USP Class L34
  • Analytical column type
  • Stainless steel hardware material
  • Ready-to-use column hardware
  • pH stability from 3.0 to 11.0
  • Ion exchange chromatography separation mode
  • HPLC technology
  • Polymer matrix
  • Spherical, non-porous particle morphology
  • Pressure stability up to 689 bar