Learn More
Waters Corp BioResolve SCX mAb Method Validation Kit, 3 μm, 2.1 mm x 100 mm, 3 pk
Catalog No. 502390456 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The BioResolve SCX mAb Method Validation Kit with VanGuard™ FIT provides reproducible, high–resolution, charged–based separations for mAbs and other proteins. It features corrosion–resistant columns with non–porous, polymer–based particles and sulfonic acid ligands, suitable for LC and LC–MS applications.
  • Corrosion–resistant columns with non–porous, polymer–based particles.
  • Grafted with negatively charged sulfonic acid ligands for high–resolution separations.
  • Ideal for LC and LC–MS applications using salt or pH gradient methods.
  • Includes VanGuard™ FIT to maximize column life and preserve biomolecule resolution.
  • Contains three columns from three distinct batches for method validation.
Catalog No. 50-239-0456 Supplier Waters Corp Supplier No. 186009197
Add to Cart
Add to Cart
Request a Quote
The BioResolve SCX mAb Method Validation Kit with VanGuard™ FIT provides reproducible, high–resolution, charged–based separations for mAbs and other proteins. It features corrosion–resistant columns with non–porous, polymer–based particles and sulfonic acid ligands, suitable for LC and LC–MS applications.
  • Corrosion–resistant columns with non–porous, polymer–based particles.
  • Grafted with negatively charged sulfonic acid ligands for high–resolution separations.
  • Ideal for LC and LC–MS applications using salt or pH gradient methods.
  • Includes VanGuard™ FIT to maximize column life and preserve biomolecule resolution.
  • Contains three columns from three distinct batches for method validation.