Learn More
Waters Corp BioResolve SCX mAb Kit, 3 μm, 4.6 mm ID, 100 mm
Catalog No. 502390774 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit features corrosion-resistant columns with non-porous, polymer-based particles and negatively charged sulfonic acid ligands, enabling reproducible, high-resolution, charge-based separations of monoclonal antibodies and other proteins in LC and LC-MS. VanGuard Fully Integrated Technology (FIT) maximizes column life without degrading biomolecule component resolution.
  • Corrosion-resistant columns for reproducible, high-resolution charge-based separations of mAbs and proteins.
  • VanGuard Fully Integrated Technology (FIT) maximizes column life and prevents contaminants.
  • Method Validation Kit includes three columns and a Charge Variant Standard for performance verification.
Catalog No. 50-239-0774 Supplier Waters Corp Supplier No. 186009199
Add to Cart
Add to Cart
Request a Quote
This kit features corrosion-resistant columns with non-porous, polymer-based particles and negatively charged sulfonic acid ligands, enabling reproducible, high-resolution, charge-based separations of monoclonal antibodies and other proteins in LC and LC-MS. VanGuard Fully Integrated Technology (FIT) maximizes column life without degrading biomolecule component resolution.
  • Corrosion-resistant columns for reproducible, high-resolution charge-based separations of mAbs and proteins.
  • VanGuard Fully Integrated Technology (FIT) maximizes column life and prevents contaminants.
  • Method Validation Kit includes three columns and a Charge Variant Standard for performance verification.