Learn More
Waters Corp HPLC Column, BioResolve SCX mAb, 3 μm, 2.1 x 100 mm, MVK
Catalog No. 502392885 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The BioResolve SCX mAb column features corrosion-resistant columns with non-porous, polymer-based particles grafted with a meticulously optimized, multi-component network of negatively charged sulfonic acid ligands. This innovative column technology provides reproducible, high-resolution, charge-based separations of mAbs and other proteins in both LC and LC-MS applications using robust salt or pH gradient methods. It also includes VanGuard™ Fully Integrated Technology (FIT) which extends column life without compromising biomolecule component resolution.
  • BioResolve column
  • Cation exchange
  • 2.1 mm inner diameter
  • Reversed phase (RP) mode
  • 3.0 μm particle size
  • 3 to 11 pH range
  • 100 mm length
Catalog No. 50-239-2885 Supplier Waters Corp Supplier No. 176004373
The BioResolve SCX mAb column features corrosion-resistant columns with non-porous, polymer-based particles grafted with a meticulously optimized, multi-component network of negatively charged sulfonic acid ligands. This innovative column technology provides reproducible, high-resolution, charge-based separations of mAbs and other proteins in both LC and LC-MS applications using robust salt or pH gradient methods. It also includes VanGuard™ Fully Integrated Technology (FIT) which extends column life without compromising biomolecule component resolution.
  • BioResolve column
  • Cation exchange
  • 2.1 mm inner diameter
  • Reversed phase (RP) mode
  • 3.0 μm particle size
  • 3 to 11 pH range
  • 100 mm length