Learn More
Waters Corp Cartridge, BioResolve SCX mAb, 3 μm, 2.1 mm X 5 mm
Catalog No. NC2391706 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These VanGuard Fully Integrated Technology (FIT) Replacement Cartridges contain 3 μm particles and are designed to protect columns from performance drops due to fouling by removing particulate contaminants. They seamlessly integrate into existing workflows. This reusable solution maintains the same inner diameter and pH range as the corresponding column for chemical and mechanical stability. While not a substitute for proper sample purification, these cartridges extend column lifetimes.
  • Designed exclusively for use with 3 μm columns with included VanGuard Fit option.
  • Seamlessly integrates into existing workflow to provide enhanced protection from particulate contaminants.
  • Reusable solution for column protection.
  • Offers the same inner diameter and pH range as the corresponding column for chemical and mechanical stability.
Catalog No. NC2391706 Supplier Waters Corp Supplier No. 186009061
These VanGuard Fully Integrated Technology (FIT) Replacement Cartridges contain 3 μm particles and are designed to protect columns from performance drops due to fouling by removing particulate contaminants. They seamlessly integrate into existing workflows. This reusable solution maintains the same inner diameter and pH range as the corresponding column for chemical and mechanical stability. While not a substitute for proper sample purification, these cartridges extend column lifetimes.
  • Designed exclusively for use with 3 μm columns with included VanGuard Fit option.
  • Seamlessly integrates into existing workflow to provide enhanced protection from particulate contaminants.
  • Reusable solution for column protection.
  • Offers the same inner diameter and pH range as the corresponding column for chemical and mechanical stability.