Learn More
Waters Corp BioResolve SEC mAb Column, 200Å, 2.5 μm, 7.8 x 150 mm
Supplier: Waters Corp 176004594
The BioResolve SEC mAb Column, 7.8 x 150 mm, utilizes 2.5 μm BEH particle technology for accurate and reproducible quantitation of monoclonal antibody (mAb) monomers, aggregates, and fragments.
- Features 2.5 μm BEH particle technology.
- Tested with mAb Size Variant Standard.
- Accurate quantitation of mAb monomers, aggregates, and fragments.
- Pore size: 200 Å; Particle size: 2.5 μm.
- Dimensions: 7.8 mm ID x 150 mm Length.
- Max pressure: 4500 psi; pH range: 2.5 to 8; Temp limits: 60 °C.
- Diol (OH) bonding.
- Molecular weight range: 10,000 to 450,000.
- Protein QC tested; HPLC system compatible; eCord enabled.
- 1 unit per package.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.