Learn More
Waters Corp BioResolve SEC mAb Column, 200Å, 2.5 μm, 4.6 x 150 mm
Catalog No. 502392201 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The BioResolve SEC mAb 4.6 x 150 mm Column utilizes 2.5 μm BEH particle technology, designed for accurate and reproducible quantitation of monoclonal antibody monomers, aggregates, and fragments. It offers intrinsic chemical stability, allowing method development across a wide pH range, and is compatible with UHPLC, HPLC, and UPLC systems. It covers a molecular weight range of 10,000 to 450,000.
  • Features 2.5 μm BEH particle technology
  • Enables accurate and reproducible quantitation of monoclonal antibody monomers, aggregates, and fragments
  • QC tested for reliable performance
  • Offers intrinsic chemical stability for wide pH range method development (2.5 to 8 pH)
  • Compatible with UHPLC, HPLC, and UPLC systems
  • Supports molecular weight range of 10,000 to 450,000
  • Guard column available to extend column lifetime
Catalog No. 50-239-2201 Supplier Waters Corp Supplier No. 176004592
The BioResolve SEC mAb 4.6 x 150 mm Column utilizes 2.5 μm BEH particle technology, designed for accurate and reproducible quantitation of monoclonal antibody monomers, aggregates, and fragments. It offers intrinsic chemical stability, allowing method development across a wide pH range, and is compatible with UHPLC, HPLC, and UPLC systems. It covers a molecular weight range of 10,000 to 450,000.
  • Features 2.5 μm BEH particle technology
  • Enables accurate and reproducible quantitation of monoclonal antibody monomers, aggregates, and fragments
  • QC tested for reliable performance
  • Offers intrinsic chemical stability for wide pH range method development (2.5 to 8 pH)
  • Compatible with UHPLC, HPLC, and UPLC systems
  • Supports molecular weight range of 10,000 to 450,000
  • Guard column available to extend column lifetime