Learn More
Waters Corp Waters BioResolve SEC mAb Column, 200Å, 2.5 μm, 4.6 x 300 mm
Supplier:  Waters Corp 186009437
Encompass_Preferred
The BioResolve SEC mAb Column is engineered for precise quantitation of monoclonal antibody monomers. It includes a size variant standard to ensure optimal LC-MS system performance for mAb analysis. This column is versatile, compatible with HPLC, UPLC, and UHPLC systems.
  • Designed for quantitation of monoclonal antibody monomers.
  • Includes a size variant standard for LC-MS system verification.
  • Compatible with HPLC, UPLC, and UHPLC systems.
  • Undergoes strict quality control testing before delivery.
  • Guard column available to extend column lifetime by trapping particulates.
  • Addresses analytical challenges in therapeutic drug analysis with multiple chemistries.
Catalog No. 50-203-9055