Learn More
Waters Corp Waters BioResolve SEC mAb Column, 200Å, 2.5 μm, 4.6 x 150 mm
Catalog No. 502039054 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This BioResolve SEC mAb Column ensures accurate and reproducible quantitation of monoclonal antibody monomers from both high and lower molecular weight fragments. It undergoes a strict QC process for batch-to-batch consistency and proper functioning. It is LC-MS system agnostic, compatible with HPLC, UHPLC, and UPLC systems, and offers a wide pH range for various analytes.
  • Ensures accurate and reproducible quantitation of mAb monomers from high and lower molecular weight fragments.
  • Strict QC process for batch-to-batch consistency and proper functioning.
  • LC-MS system agnostic, compatible with HPLC, UHPLC, and UPLC systems.
  • Includes an mAb size variant standard to verify LC-MS instrument performance.
  • Offers a wide pH range for various analytes.
  • Can be protected from contaminants using an appropriate guard column.
  • Maximum pressure limit is 4,500 psi (310 bar).
Catalog No. 50-203-9054 Supplier Waters Corp Supplier No. 186009435
This BioResolve SEC mAb Column ensures accurate and reproducible quantitation of monoclonal antibody monomers from both high and lower molecular weight fragments. It undergoes a strict QC process for batch-to-batch consistency and proper functioning. It is LC-MS system agnostic, compatible with HPLC, UHPLC, and UPLC systems, and offers a wide pH range for various analytes.
  • Ensures accurate and reproducible quantitation of mAb monomers from high and lower molecular weight fragments.
  • Strict QC process for batch-to-batch consistency and proper functioning.
  • LC-MS system agnostic, compatible with HPLC, UHPLC, and UPLC systems.
  • Includes an mAb size variant standard to verify LC-MS instrument performance.
  • Offers a wide pH range for various analytes.
  • Can be protected from contaminants using an appropriate guard column.
  • Maximum pressure limit is 4,500 psi (310 bar).