Learn More
Waters Corp WATERS CORPORATION BioResolve SEC mAb Column, 200 Å, 2.5 μm, 7.8 x 300 mm
Supplier:  Waters Corp 186009441
Encompass_Preferred
The BioResolve SEC mAb Column, 200 Å, 2.5 μm, 7.8 x 300 mm, is designed for Size Exclusion Chromatography (SEC) applications. It is specifically used for the analysis of monoclonal antibodies (mAbs) and their size variants, such as high molecular weight species (HMWS) or aggregates. This column improves resolution, especially for low molecular weight species (LMWS), enabling more accurate quantitation.
  • Resolves the LMWS 1 fragment that co-elutes with the monomer peak when using conventional SEC columns.
  • Enables more accurate quantitation of the monomer peak by resolving the back-shoulder peak.
  • Offers greater efficiency, selectivity, and enhanced resolution when combined with modern UHPLC systems.
Catalog No. 50-203-9057