Learn More
Waters Corp Waters SEC mAb Column, 200Å, 2.5 μm, 7.8 × 300 mm
Catalog No. 502037643 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This SEC mAb column is designed for the separation of monoclonal antibodies and their size variants. It is suitable for evaluating SEC-MALS data quality and for molecular weight determination of substances like trastuzumab. The column provides excellent signal-to-noise ratio in MALS detection and delivers consistent results for molecular weight analysis.
  • Designed for separation of monoclonal antibodies and their size variants.
  • Suitable for evaluating SEC-MALS data quality.
  • Used for molecular weight determination.
  • Provides 1.5–2 times higher signal-to-noise ratio in MALS detection compared to other columns.
  • Features significantly lower MALS noise than diol-bonded silica-based SEC columns.
  • Delivers consistent results for molecular weight of monomer and HMWS1.
  • Supplied with a mAb size variant standard.
Catalog No. 50-203-7643 Supplier Waters Corp Supplier No. 176004595
This SEC mAb column is designed for the separation of monoclonal antibodies and their size variants. It is suitable for evaluating SEC-MALS data quality and for molecular weight determination of substances like trastuzumab. The column provides excellent signal-to-noise ratio in MALS detection and delivers consistent results for molecular weight analysis.
  • Designed for separation of monoclonal antibodies and their size variants.
  • Suitable for evaluating SEC-MALS data quality.
  • Used for molecular weight determination.
  • Provides 1.5–2 times higher signal-to-noise ratio in MALS detection compared to other columns.
  • Features significantly lower MALS noise than diol-bonded silica-based SEC columns.
  • Delivers consistent results for molecular weight of monomer and HMWS1.
  • Supplied with a mAb size variant standard.