Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Column, SEC, 200 Å, 2.5 μm, 7.8 x 300 mm

Catalog No. 502391291 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The BioResolve SEC mAb 7.8 x 300 mm Column and Guard is designed for reproducible and accurate quantitation of monoclonal antibodies (mAb) from frequently associated high molecular weight aggregates and lower molecular weight fragments. Featuring 2.5 μm BEH particle technology, it provides flexibility for both HPLC and UPLC systems. This column set is manufactured under cGMP and ISO 9001 certified conditions and undergoes rigorous QC testing to ensure consistent performance.

  • Application specifically tested with mAb Size Variant Standard.
  • mAb Size Variant Standard included.
  • eCord enabled.
  • Chemistry: SEC.
  • Particle substrate: Hybrid.
  • Bonding technology: Diol (OH).
  • Particle shape: Spherical.
  • Endfitting type: Parker-style.
  • Pore size: 200 Å.
  • Dimensions: 7.8 mm ID x 300 mm length.

Catalog No. 50-239-1291 Supplier Waters Corp Supplier No. 176004599
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The BioResolve SEC mAb 7.8 x 300 mm Column and Guard is designed for reproducible and accurate quantitation of monoclonal antibodies (mAb) from frequently associated high molecular weight aggregates and lower molecular weight fragments. Featuring 2.5 μm BEH particle technology, it provides flexibility for both HPLC and UPLC systems. This column set is manufactured under cGMP and ISO 9001 certified conditions and undergoes rigorous QC testing to ensure consistent performance.

  • Application specifically tested with mAb Size Variant Standard.
  • mAb Size Variant Standard included.
  • eCord enabled.
  • Chemistry: SEC.
  • Particle substrate: Hybrid.
  • Bonding technology: Diol (OH).
  • Particle shape: Spherical.
  • Endfitting type: Parker-style.
  • Pore size: 200 Å.
  • Dimensions: 7.8 mm ID x 300 mm length.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.