Learn More
Waters Corp BioResolve SEC mAb Column, 200Å, 2.5 μm, 7.8 x 150 mm
Catalog No. 502391859 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This SEC mAb Column, with an integrated SEC mAb Guard, is designed to support legacy methods for the accurate and reproducible quantitation of monoclonal antibody monomers from high molecular weight aggregates and lower molecular weight fragments. It features 2.5 μm BEH particle technology, is operable up to 60°C, and includes a guard column to protect against mobile phase contaminants, ensuring extended column lifetime.
  • Supports legacy methods for monoclonal antibody analysis.
  • Utilizes 2.5 μm BEH particle technology for high-resolution separations.
  • Tested for accurate quantitation of monoclonal antibody monomers, aggregates, and fragments.
  • Includes a guard column for protection against mobile phase contaminants.
  • Offers a wide pH range of 2.5 to 8.
  • Maintains mechanical stability under high backpressures.
  • Designed for HPLC systems with SEC chemistry.
Catalog No. 50-239-1859 Supplier Waters Corp Supplier No. 176004598
Add to Cart
Add to Cart
This SEC mAb Column, with an integrated SEC mAb Guard, is designed to support legacy methods for the accurate and reproducible quantitation of monoclonal antibody monomers from high molecular weight aggregates and lower molecular weight fragments. It features 2.5 μm BEH particle technology, is operable up to 60°C, and includes a guard column to protect against mobile phase contaminants, ensuring extended column lifetime.
  • Supports legacy methods for monoclonal antibody analysis.
  • Utilizes 2.5 μm BEH particle technology for high-resolution separations.
  • Tested for accurate quantitation of monoclonal antibody monomers, aggregates, and fragments.
  • Includes a guard column for protection against mobile phase contaminants.
  • Offers a wide pH range of 2.5 to 8.
  • Maintains mechanical stability under high backpressures.
  • Designed for HPLC systems with SEC chemistry.