Learn More
Waters Corp Waters Corporation SEC mAb Column, 200Å, 2.5 μm, 4.6 x 300 mm
Supplier:  Waters Corp 176004597
Encompass_Preferred
This SEC mAb column and guard combination utilizes 2.5 μm BEH particle technology for accurate and reproducible quantitation of monoclonal antibody monomers, aggregates, and fragments. It is compatible with UHPLC, HPLC, and UPLC systems, and each unit undergoes a strict QC process.
  • Features 2.5 μm BEH particle technology for superior separation performance and stability
  • Specifically tested with the mAb size variant standard for reliable analysis
  • Rigid BEH particle enables a wider, more usable pH range for method development
  • LC system agnostic, compatible with UHPLC, HPLC, and UPLC systems
  • Diol (OH) bonding technology
  • Includes an accompanying guard column for protection against contaminants
Catalog No. 50-203-7645