Learn More
Waters Corp Waters Corporation BioResolve RP mAb Polyphenyl Column, 450Å, 2.7 μm, 4.6 mm x 150 mm
Supplier:  Waters Corp 176004169
Encompass_Preferred
The BioResolve RP mAb Polyphenyl Column analyzes intact IgG mAbs & fragments by LC or LC-MS, featuring 2.7-μm superficially-porous-silica particles with 450Å pore diameter & proprietary polyphenyl bonding.
  • For LC or LC-MS analysis of intact IgG mAbs and fragments.
  • Includes Intact mAb Mass Check and mAb Subunit Standards.
  • Improved peak capacity for better resolution.
  • Minimal injection-to-injection carryover for confident analyses.
  • Compatible with UHPLC, UPLC, and HPLC systems.
  • Novel phenyl bonding technology for higher flow rates.
  • Maximizes resolving power and protein selectivity.
  • High-coverage bonding for extended column lifetimes and low pH stability.
  • Strict QC process, tested with mAb Subunit Standard.
  • pH range of 2 to 7.
Catalog No. 50-203-7571