Learn More
Research Products International Corp BioSafe Barrier Serological Pipets, 25ml
SDP
Catalog No. 501977549
Encompass
BioSafe Pipets feature a unique barrier filter that "locks" liquid in pipet during over aspiration. Ideal pipets provide sample integrity, eliminate aerosols, and provide total contamination protection.
Catalog No. 50-197-7549 Supplier Research Products International Corp Supplier No. 248617
BioSafe Pipets feature a unique barrier filter that "locks" liquid in pipet during over aspiration. Ideal pipets provide sample integrity, eliminate aerosols, and provide total contamination protection.