Learn More
Research Products International Corp BioSafe Barrier Serological Pipets, 5 ml, 0.1 ml Graduations, 200 per Case
SDP
Catalog No. 501367564
Encompass
BioSafe Pipets feature a unique barrier filter that "locks" liquid in pipet during over aspiration. Ideal pipets provide sample integrity, eliminate aerosols, and provide total contamination protection.
Catalog No. 50-136-7564 Supplier Research Products International Corp Supplier No. 248615
BioSafe Pipets feature a unique barrier filter that "locks" liquid in pipet during over aspiration. Ideal pipets provide sample integrity, eliminate aerosols, and provide total contamination protection.