Learn More
Research Products International Corp BioSafe Barrier Serological Pipets, 10ml
SDP
Catalog No. 501977829
Encompass
BioSafe Pipets feature a unique barrier filter that "locks" liquid in pipet during over aspiration. Ideal pipets provide sample integrity, eliminate aerosols, and provide total contamination protection.
Catalog No. 50-197-7829 Supplier Research Products International Corp Supplier No. 248616
BioSafe Pipets feature a unique barrier filter that "locks" liquid in pipet during over aspiration. Ideal pipets provide sample integrity, eliminate aerosols, and provide total contamination protection.