Learn More
Research Products International Corp Bio-Safe NA Complete Counting Cocktail, 4 Liters, 4 Bottles per Case
SDP
Catalog No. 50476611
Encompass_Preferred
Bio-Safe NA provides the highest efficiency for counting organic samples, dry filters and samples treated with tissue solubilizers such as TS-2. Effective and economical cocktail for counting dry wipe tests in radiation safety procedures. Compatible with all plastic scintillation vials.
Catalog No. 50-476-611 Supplier Research Products International Corp Supplier No. 111198CS
Bio-Safe NA provides the highest efficiency for counting organic samples, dry filters and samples treated with tissue solubilizers such as TS-2. Effective and economical cocktail for counting dry wipe tests in radiation safety procedures. Compatible with all plastic scintillation vials.