Learn More
Air Science Usa Biosafety Safety 72" / 1800mm Nominal Width, NSF Listed, Biological Safety Cabinet
SDP
Catalog No. NC2684366
Encompass_Preferred
Purair Bio Class II A2 Biological Safety Cabinet 72" / 1800mm Nominal Width, NSF Listed, 115V 60Hz, Includes: Adjustable Height Base Stand on Levelers, UV system, Two GFI Outlets, One Service Fixture
Catalog No. NC2684366 Supplier Air Science Usa Supplier No. ASAHA193CAA
Add to Cart
Add to Cart
Request a Quote
Purair Bio Class II A2 Biological Safety Cabinet 72" / 1800mm Nominal Width, NSF Listed, 115V 60Hz, Includes: Adjustable Height Base Stand on Levelers, UV system, Two GFI Outlets, One Service Fixture