Learn More
Description
Specifications
Specifications
| Certifications/Compliance | LUO, MUO |
| Content And Storage | Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Format | Bottle |
| For Use With (Application) | Nutritional supplement in vitamin assays, susceptibility testing, and other laboratory media, and microbial fermentation in which the high salt content will not interfere |
| Product Line | Biosate |
| Quantity | 454 g |
| Source | Animal origin, casein and yeast |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
