Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BIOShield™ 1000A Rotor Adapters
GSA_VA

Catalog No. 75003642
Change view
Click to view available options
Capacity:
15 mL Conical Tube
50 mL Conical Tube
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
75-003-642 15 mL Conical Tube
75-003-643 50 mL Conical Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 75-003-642 Supplier Thermo Scientific™ Supplier No. 75003642
Add to Cart
Add to Cart
This item is not returnable. View return policy

Increase rotor versatility with Thermo Scientific™ BIOShield™ 1000A Rotor Adapters, which provide support for 15/50mL conical tubes.

Ordering Information:

A variety of other adapters (sold in sets of four unless otherwise noted) are available for use with both the BIOShield 1000A Rotor (75-003-603) and the TX-750 Swinging-Bucket Rotor with Rectangular Buckets (75-003-607/75-003-614). Contact your sales representative for more information.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Type Rotor Tube Adapter
Capacity 15 mL Conical Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.