Learn More
ILC Dover BioShield™ Full Hood
Catalog No. 19001504
Change view
Click to view available options
Quantity:
25/Pk.
3/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-001-504 3/Pk.
19-001-505 25/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-001-504 Supplier ILC Dover Supplier No. S20283
Add to Cart
Add to Cart
Designed in collaboration with leading healthcare organizations, this loose-fitting PAPR with an APF of 1,000 is made from spun bonded polyolefin, coated, and latex free.
  • Visor is impact resistant, made with optical grade polycarbonate
  • Internal and external skirt interfaces with most protective suits and requires no fit testing
Specifications
Material Polycarbonate, Polyolefin
Latex Free Yes
Quantity 3/Pk.