Learn More
AirSea USA UN3373 Packaging – Bioshipper-2 is suitable for Biological Substances, Category B UN3373
Catalog No. NC2101122
Encompass_Preferred
The Air Sea Containers BioShipper-2 is supplied with 1 x 95KPa BioBag-2, 1 x Security Seal 2, 2 x 4 Bay Absorbent Cushioning Pouch and Assembly Sheet, detailing how substances should be transported inside. For higher volume shipments using our 95KPa BioBag-2 this kit can carry up to 2 x cryovial boxes or a multitude of single primary containers wrapped in absorbent cushioning. The BioShipper-2 is designed to be cost efficient and is delivered flat packed with fast easy pop-up assembly.
This item is not returnable. View return policy
Catalog No. NC2101122 Supplier AirSea USA Supplier No. 762
The Air Sea Containers BioShipper-2 is supplied with 1 x 95KPa BioBag-2, 1 x Security Seal 2, 2 x 4 Bay Absorbent Cushioning Pouch and Assembly Sheet, detailing how substances should be transported inside. For higher volume shipments using our 95KPa BioBag-2 this kit can carry up to 2 x cryovial boxes or a multitude of single primary containers wrapped in absorbent cushioning. The BioShipper-2 is designed to be cost efficient and is delivered flat packed with fast easy pop-up assembly.