Learn More
Abbott™ Biosite™ Triage™ Drugs of Abuse Panel
Catalog No. 23034973
Change view
Click to view available options
Quantity:
25 Tests
Type:
Triage™ Drugs of Abuse Panel plus TCA
Catalog No. Quantity Type
23-034-973 25 Tests Triage™ Drugs of Abuse Panel plus TCA
1 options
Catalog No. 23-034-973 Supplier Abbott™ Supplier No. 92000
Immunoassay for the qualitative determination of the presence of parent compound and/or major metabolites of up to nine distinct drug classes in urine
  • Test results from a urine sample in about 10 minutes
  • CLIA Complexity: Moderate
  • A self-contained, disposable, credit card-sized test device
  • Easy-to-read results for up to nine drug classes
  • Tests include amphetamines/methamphetamines, barbiturates, benzodiazepines, cocaine, marijuana, methadone, opiates, PCP and tricyclic antidepressants
  • The presence of color in any drug detection zone indicates the presence of the drug at or above the threshold concentrations
  • Built-in positive and negative controls in every device validate results
  • Kit sizes available in 25 test configurations
Specifications
Clia Complexity Moderate
Sample Type Urine
Type Triage™ Drugs of Abuse Panel plus TCA
Includes Amphetamines, Methamphetamines, Barbiturates, Benzodiazepines, Cocaine, Marijuana, Methadone, Opiates, PCP, Propoxyphene and Tricyclic Antidepressants
Quantity 25 Tests
Test Time 10 min.