Learn More
Sartorius Sartorius 18-5117 High Precision SAX Biosensors, CV <4%, 96 Biosensors per Tray
Supplier:  Sartorius 185117
Encompass
The Sartorius 18-5117 Octet High Precision Streptavidin (SAX) Biosensors are designed for advanced bio-layer interferometry (BLI) applications. They offer exceptional precision with a coefficient of variation (CV) of less than 4%, ensuring accurate quantitation, screening, and kinetic analysis. Supplied in trays of 96, these ready-to-use biosensors facilitate high-throughput experiments, setting them apart from standard options in the market.
Catalog No. NC0784278
Add to Cart
Description
Description
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov