Learn More
BioSpec Products BioSpec Products Mini-BeadBeater-16 ULTRA, Blue, 120v, Beadbeater Homogenizer, Sixteen 2ml tube capacity
SDP
Catalog No. NC3973096
Encompass_Preferred
The Mini-Beadbeater-16 Ultra is a high-performance, multi-sample homogenizer designed for rapid and efficient disruption of biological samples. BioSpec’s next-generation bead beater processes up to 16 microvials simultaneously using a high-speed, near-horizontal shaking motion that maximizes bead impact for superior lysis. Tough samples such as microbial cells, plant tissues, and fibrous materials are homogenized in minutes.Samples are processed in sealed, disposable microvials to prevent cross-contamination while preserving sensitive biomolecules including DNA, RNA, and proteins. With a powerful motor and consistent high-speed operation, the system requires no cool-down time between runs, enabling continuous use.Ideal for PCR, sequencing, and protein analysis, the Mini-Beadbeater-16 Ultra delivers reliable, high-yield results in a compact, easy-to-use format designed for modern, shared labs.
Catalog No. NC3973096 Supplier BioSpec Products Supplier No. 6202101BL
The Mini-Beadbeater-16 Ultra is a high-performance, multi-sample homogenizer designed for rapid and efficient disruption of biological samples. BioSpec’s next-generation bead beater processes up to 16 microvials simultaneously using a high-speed, near-horizontal shaking motion that maximizes bead impact for superior lysis. Tough samples such as microbial cells, plant tissues, and fibrous materials are homogenized in minutes.Samples are processed in sealed, disposable microvials to prevent cross-contamination while preserving sensitive biomolecules including DNA, RNA, and proteins. With a powerful motor and consistent high-speed operation, the system requires no cool-down time between runs, enabling continuous use.Ideal for PCR, sequencing, and protein analysis, the Mini-Beadbeater-16 Ultra delivers reliable, high-yield results in a compact, easy-to-use format designed for modern, shared labs.