Learn More
BioSpec Products BioSpec Products Mini-BeadBeater-16 ULTRA, Red, 115V
SDP
Catalog No. NC3973097
Encompass_Preferred
The Mini-BeadBeater-16 ULTRA is BioSpec's next-generation laboratory bead mill designed for rapid, reliable disruption of microbial cells and plant or animal tissues. It provides powerful homogenization with enhanced safety, quieter operation, and a modern design, all within a compact benchtop footprint.
  • High-performance bead mill homogenizer
  • Rapid, reliable disruption of microbial cells and plant or animal tissues
  • Enhanced safety
  • Quieter operation
  • Modern design
  • Compact bench top footprint
Catalog No. NC3973097 Supplier BioSpec Products Supplier No. 6202101RD
The Mini-BeadBeater-16 ULTRA is BioSpec's next-generation laboratory bead mill designed for rapid, reliable disruption of microbial cells and plant or animal tissues. It provides powerful homogenization with enhanced safety, quieter operation, and a modern design, all within a compact benchtop footprint.
  • High-performance bead mill homogenizer
  • Rapid, reliable disruption of microbial cells and plant or animal tissues
  • Enhanced safety
  • Quieter operation
  • Modern design
  • Compact bench top footprint