Learn More
Bioworld BIOSPECTRUM DYE-AFFINITY CO
Catalog No. 50197893
Encompass_Preferred
BIOSPECTRUM DYE-AFFINITY COLUMN SET: It is a rapid, inexpensive, and versatile method that is applicable to the purification of many proteins and to demonstrate principles of bio affinity chromatography. Dye chromatography use mimics of...
Catalog No. 50-197-893 Supplier Bioworld Supplier No. 201810871
BIOSPECTRUM DYE-AFFINITY COLUMN SET: It is a rapid, inexpensive, and versatile method that is applicable to the purification of many proteins and to demonstrate principles of bio affinity chromatography. Dye chromatography use mimics of...