Learn More
INDICAL INC BIOSPRINT 96 ONE-ALL VET KIT
SDP
Catalog No. NC3343880
Encompass_Preferred
The BioSprint 96 One-For-All Vet Kit offers an automated solution for the purification of viral RNA/DNA and bacterial DNA from a wide range of samples. Designed for use with 96-well magnetic particle processors, this kit streamlines the nucleic acid extraction process, enabling high-throughput processing with minimal hands-on time.The all-in-one kit includes all necessary reagents and 96-well plasticware, providing a convenient, ready-to-use solution for laboratories. This automated procedure ensures reliable and efficient results, saving valuable time and reducing the potential for user error. Ideal for veterinary applications, it simplifies the workflow for viral, bacterial, and other pathogen detection across various sample types.
Catalog No. NC3343880 Supplier INDICAL INC Supplier No. SP947057
The BioSprint 96 One-For-All Vet Kit offers an automated solution for the purification of viral RNA/DNA and bacterial DNA from a wide range of samples. Designed for use with 96-well magnetic particle processors, this kit streamlines the nucleic acid extraction process, enabling high-throughput processing with minimal hands-on time.The all-in-one kit includes all necessary reagents and 96-well plasticware, providing a convenient, ready-to-use solution for laboratories. This automated procedure ensures reliable and efficient results, saving valuable time and reducing the potential for user error. Ideal for veterinary applications, it simplifies the workflow for viral, bacterial, and other pathogen detection across various sample types.