Learn More
BioSpec Products BioSqueezer, quick freeze 10-100 mg of soft animal and human tissue.
SDP
Catalog No. NC1033496
Encompass_Preferred
The BioSqueezer is used to "snap-freeze" (quickly freeze) 10-100 mg of soft animal and human tissue. It does the same thing as a classic freeze clamp but is easier to use and gives more reproducable results.Fresh tissue is compressed into a thin disc about the size of a small coin under pressure from a stainless steel clamp head that has been pre-cooled in liquid nitrogen or dry ice. Freezing occurs in a few milliseconds, thus offering a "snapshot" of metabolic events in the cells. The hard frozen disc is easily fragmented by hand or by grounding in a Cryo-Cup mortar and pestle for subsequent rapid extraction of labile nucleic acids, proteins and other intracellular products.
Catalog No. NC1033496 Supplier BioSpec Products Supplier No. 1210
The BioSqueezer is used to "snap-freeze" (quickly freeze) 10-100 mg of soft animal and human tissue. It does the same thing as a classic freeze clamp but is easier to use and gives more reproducable results.Fresh tissue is compressed into a thin disc about the size of a small coin under pressure from a stainless steel clamp head that has been pre-cooled in liquid nitrogen or dry ice. Freezing occurs in a few milliseconds, thus offering a "snapshot" of metabolic events in the cells. The hard frozen disc is easily fragmented by hand or by grounding in a Cryo-Cup mortar and pestle for subsequent rapid extraction of labile nucleic acids, proteins and other intracellular products.