Learn More
AdipoGen BioStab Antibody Stabilizer
SDP
Catalog No. 502563396
Encompass_Preferred
Free or conjugated antibodies are often used in immunoassays, either in the system specific detector antibody plus streptavidin-HRP or in the system analyte-specific antibody plus species-specific antibody-HRP-conjugate. Both systems allow a high flexibility at lower costs. BioStab Antibody Stabilizer dramatically extends the life of your valuable antibodies, allowing much more consistent results over extended periods of time compared to daily prepared dilutions. BioStab Antibody Stabilizer can be used in ELISA, Western Blotting, Immunohistochemistry and any other applications which require marker or biotinylated antibodies. It is supplied as a ready-to-use solution for dilution of antibodies. This stabilizer contains a minimum amount of a surfactant. The stabilizer is free of any protein to minimize potential unspecific interactions. The method below is strongly recommended for standard assay procedures and reduces loss of valuable conjugates and assay-to-assay variations.
Catalog No. 50-256-3396 Supplier AdipoGen Supplier No. AG10T0031L250
Free or conjugated antibodies are often used in immunoassays, either in the system specific detector antibody plus streptavidin-HRP or in the system analyte-specific antibody plus species-specific antibody-HRP-conjugate. Both systems allow a high flexibility at lower costs. BioStab Antibody Stabilizer dramatically extends the life of your valuable antibodies, allowing much more consistent results over extended periods of time compared to daily prepared dilutions. BioStab Antibody Stabilizer can be used in ELISA, Western Blotting, Immunohistochemistry and any other applications which require marker or biotinylated antibodies. It is supplied as a ready-to-use solution for dilution of antibodies. This stabilizer contains a minimum amount of a surfactant. The stabilizer is free of any protein to minimize potential unspecific interactions. The method below is strongly recommended for standard assay procedures and reduces loss of valuable conjugates and assay-to-assay variations.