Learn More
AdipoGen BioStab PCR Optimizer
SDP
Catalog No. 502563393
Encompass_Preferred
PCR is a technique extensively used in different applications in molecular biology, molecular diagnostics and forensics. However, some sequences, for example the 5´-regions of many eukaryotic genes, are difficult to amplify. Incomplete separation of DNA strands results in little or no yield of desired product and multiple non-specific PCR-fragments. BioStab PCR Optimizer solves this problem and outperforms betaine. The mode of action of BioStab PCR optimizer comprises lowering the melting temperature of GC pairs without affecting that of AT pairs. The BioStab PCR optimizer is designed as a PCR additive which improves the yield and specificity of PCR reaction and allows amplification of difficult targets e.g. sequences with >70% GC pairs. BioStab PCR optimizer enables the use of a wide range of annealing temperatures thereby eliminating the need for an optimization of amplification conditions. The BioStab PCR optimizer is provided as 5x concentrated solution.
Catalog No. 50-256-3393 Supplier AdipoGen Supplier No. AG10T0032L005
PCR is a technique extensively used in different applications in molecular biology, molecular diagnostics and forensics. However, some sequences, for example the 5´-regions of many eukaryotic genes, are difficult to amplify. Incomplete separation of DNA strands results in little or no yield of desired product and multiple non-specific PCR-fragments. BioStab PCR Optimizer solves this problem and outperforms betaine. The mode of action of BioStab PCR optimizer comprises lowering the melting temperature of GC pairs without affecting that of AT pairs. The BioStab PCR optimizer is designed as a PCR additive which improves the yield and specificity of PCR reaction and allows amplification of difficult targets e.g. sequences with >70% GC pairs. BioStab PCR optimizer enables the use of a wide range of annealing temperatures thereby eliminating the need for an optimization of amplification conditions. The BioStab PCR optimizer is provided as 5x concentrated solution.