Learn More
AdipoGen BioStab Peroxidase Conj Stabil
SDP
Catalog No. 502563398
Encompass_Preferred
Horseradish peroxidase HRP is an enzyme commonly used for immunodetection. BioStab Peroxidase Conjugate Stabilizer extends the lifetime of HRP-antibody conjugates and increases the signal intensity, allowing much more consistent results over extended periods of time compared to daily prepared dilutions. BioStab Peroxidase Conjugate Stabilizer can be used in ELISA, Western Blotting, Immunohistochemistry and any other applications which require HRP conjugates. It is supplied as ready-to-use solution for dilution of HRP conjugates. This stabilizer contains a minimum amount of a surfactant and is free of any protein to minimize potential unspecific interactions.
Catalog No. 50-256-3398 Supplier AdipoGen Supplier No. AG10T0030L250
Horseradish peroxidase HRP is an enzyme commonly used for immunodetection. BioStab Peroxidase Conjugate Stabilizer extends the lifetime of HRP-antibody conjugates and increases the signal intensity, allowing much more consistent results over extended periods of time compared to daily prepared dilutions. BioStab Peroxidase Conjugate Stabilizer can be used in ELISA, Western Blotting, Immunohistochemistry and any other applications which require HRP conjugates. It is supplied as ready-to-use solution for dilution of HRP conjugates. This stabilizer contains a minimum amount of a surfactant and is free of any protein to minimize potential unspecific interactions.