Learn More
Chemglass Life Sciences PTFE Impeller Assembly, Low Profile, 171mm OAW x 162mm OAH, for 3L VSA
SDP
Catalog No. 501532231
Encompass
Our PTFE impeller assemblies are designed to provide agitation forvertical side arm (VSA) vessels and spinners when used withcap adapter and Dura Drive overhead motor.PLEASE NOTE: STAINLESS STEEL IMPELLER ASSEMBLIES ARE AVAILABLE AS SPECIAL ORDER.
Catalog No. 50-153-2231 Supplier Chemglass Life Sciences Supplier No. CLS144044
Our PTFE impeller assemblies are designed to provide agitation forvertical side arm (VSA) vessels and spinners when used withcap adapter and Dura Drive overhead motor.PLEASE NOTE: STAINLESS STEEL IMPELLER ASSEMBLIES ARE AVAILABLE AS SPECIAL ORDER.