Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Diol (OH) Column, 125Å, 10 μm, 7.5 mm x 300 mm

Catalog No. 50780978 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This product primarily supports legacy methods. SEC columns are available in 4.6 mm, 7.5 mm, and 7.8 mm i.d. analytical and 21.5 mm i.d. preparative and guard columns.

  • Chemistry: Diol (OH)
  • Separation mode: SEC
  • Particle substrate: Silica
  • pH range: 2.5 to 7.5
  • Maximum pressure: 300 psi (20 bar)
  • Endcapped: Yes
  • Molecular weight range: 5000 to 150000
  • Particle shape: Spherical
  • Particle size: 10 μm
  • Endfitting type: Parker-style
  • Pore size: 125 Å
  • Format: Column
  • System: HPLC
  • USP classification: L59
  • Inner diameter: 7.5 mm
  • Length: 300 mm
  • Application: Protein
  • Units per package: 1 pk

Catalog No. 50-780-978 Supplier Waters Corp Supplier No. 186002168
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product primarily supports legacy methods. SEC columns are available in 4.6 mm, 7.5 mm, and 7.8 mm i.d. analytical and 21.5 mm i.d. preparative and guard columns.

  • Chemistry: Diol (OH)
  • Separation mode: SEC
  • Particle substrate: Silica
  • pH range: 2.5 to 7.5
  • Maximum pressure: 300 psi (20 bar)
  • Endcapped: Yes
  • Molecular weight range: 5000 to 150000
  • Particle shape: Spherical
  • Particle size: 10 μm
  • Endfitting type: Parker-style
  • Pore size: 125 Å
  • Format: Column
  • System: HPLC
  • USP classification: L59
  • Inner diameter: 7.5 mm
  • Length: 300 mm
  • Application: Protein
  • Units per package: 1 pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.